Learn More
Invitrogen™ Mouse TNF alpha ELISA Kits
Description
Assay TNFα (mouse tumor necrosis factor alpha) levels in mouse serum and culture supernatant samples with our Thermo Scientific™ Mouse TNF alpha ELISA Kits.
Target: Natural and recombinant mouse TNFα (mouse tumor necrosis factor alpha)
Format: Colorimetric ELISA (TMB substrate) in clear 96 well strip plate
Measurement: Absorbance at 450nm minus absorbance at 550nm
Assay Range: 50 to 2450pg/mL
Sensitivity: < 9pg/mL
Precision: Intra-assay CV < 10%; inter-assay CV < 10%
Sample Types: Cell culture supernatant, serum
Total Assay Time: 3 hours
Sample Size: 50μL
Specificity: No significant cross-reaction with mouse IL-2, IL-3, IL-4, IL-5, IL-6, GM-CSF, IFNα or human TNFα. This ELISA has some cross-reactivity with rabbit and cynomologous monkey TNFα in culture supernatants
Includes:Complete Kits contain pre-coated antibody-plates, detection antibodies, buffers, diluents, standards, and substrates. The Antibody Pair Kit (previously called a “MiniKit”) contains only matched antibodies and standard (no plates or detection reagents).
Specifications
Specifications
| Assay Range | 50 to 2450pg/mL |
| Assay Sensitivity | <9pg/mL |
| Description | Mouse TNF alpha ELISA Kit |
| Immunoassay Kit Format | Colorimetric ELISA (TMB substrate) with Plates |
| Sample Type | Cell Culture Supernatant, Serum |
| Specificity | No significant cross-reaction with mouse IL-2, IL-3, IL-4, IL-5, IL-6, GM-CSF, IFNa or human TNFa. This ELISA has some cross-reactivity with rabbit and cynomologous monkey TNFa in culture supernatants |
| Sufficient For | 5 x 96 microplate wells |
| Kit Contents | Streptavidin-HRP Concentrate, 250 μL, TMB Substrate Solution, 5 x 13mL, Stop Solution, 55mL, Standard Diluent, 55mL, Lyophilized Recombinant Mouse TNF alpha Standard, 5 vials, HRP Dilution Buffer, 70mL, Biotinylated Antibody Reagent, 30mL, Anti-Mouse TNF alpha Precoated 96-well Strip Plate, 5 ea, Adhesive Plate Covers, 30 ea, 30X Wash Buffer, 200mL |
| Measurement | Absorbance at 450nm minus absorbance at 550nm |
| Precision | Intra-assay CV <10%; inter-assay CV <10% |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.