Learn More
STEMCELL Technologies Mouse TNF-alpha ELISA Kit
Product Code. 17138041
Encompass_Preferred
Click to view available options
Quantity:
2 Plates
Unit Size:
Pack of 2
Product Code. 17138041
Brand: STEMCELL Technologies 02030
For detection and measurement of mouse tumor necrosis factor-alpha
  • The Mouse Tumor Necrosis Factor-alpha (TNF-α) ELISA Kit is designed for the quantitative detection and measurement of mouse TNF-α in biological fluids such as serum, plasma, and cell culture supernatants
  • TNF-α is produced by activated T cells and macrophages in response to inflammation or infection
  • It is also produced by B cells, NK cells, mast cells, neutrophils, dendritic cells, microglia, endothelial cells, smooth muscle cells, cardiomyocytes, and fibroblasts
  • The assay is based on the sandwich ELISA method, in which samples are added to ELISA strip plates pre-coated with capture antibodies specific for the cytokine
  • The captured cytokine is detected by addition of a biotinylated detection antibody, followed by streptavidin-horseradish peroxidase, which binds the biotinylated antibody
  • Addition of the chromogenic enzyme substrate 3,3',5,5' tetramethylbenzidine (TMB) results in a colored product with an intensity directly proportional to the concentration of cytokine in the sample
Specifications
Description B Cells; Cardiomyocytes, PSC-Derived; Dendritic Cells; Endothelial Cells; Granulocytes and Subsets; Macrophages; Mouse Embryonic Fibroblasts; Myogenic Stem and Progenitor Cells; NK Cells; T Cells
For Use With (Application) ELISA
Quantity 2 Plates