missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Mouse TMEM119 (NP_666274) VersaClone cDNA
Description
- Also known as: obif; AW208946; BC025600
- TMEM119 is a single span type I membrane protein and is enriched in microglia.
- Microglia are myeloid cells of the central nervous system (CNS) that participate both in normal CNS function and disease.
- TMEM119 plays an important role in bone formation and normal bone mineralization and promotes the differentiation of myoblasts into osteoblasts.
- The insert is sequence verified and the entire plasmid DNA and insert translated sequences are provided.
- For reasearch use only. Not for use in humans.
Specifications
Specifications
| Gene | mTmem119 |
| Accession Number | NP_666274 |
| Concentration | 10 μg at 0.2 μg/μL |
| Formulation | cDNA is provided in 10 mM Tris-Cl, pH 8.5 |
| Insert Type | 856 bp |
| Gene ID (Entrez) | 338773 |
| Content And Storage | Store the unopened product at -20°C to -70°C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date. |
| Species | Mouse |
| Quantity | 10 μg |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?