missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse TLR6 Antibody, R&D Systems™

Rat Monoclonal Antibody
Brand: R&D Systems MAB1533-SP
This item is not returnable.
View return policy
Description
TLR6 Monoclonal specifically detects TLR6 in Mouse samples. It is validated for Flow Cytometry, CyTOF-ready.
Specifications
| TLR6 | |
| Monoclonal | |
| LYOPH | |
| Flow Cytometry 0.25 ug/10^6 cells, CyTOF-ready | |
| BAA78632 | |
| TLR6 | |
| HEK293 human embryonic kidney cell line transfected with mouse TLR6 Phe39-Thr806 Accession # BAA78632 | |
| 25 μg | |
| Primary | |
| Detects mouse TLR6. Stains mouse TLR6 transfectants and not irrelevant transfectants. | |
| Mouse | |
| Purified |
| Flow Cytometry, CyTOF | |
| 418601 | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| CD286, CD286 antigen, toll-like receptor 6 | |
| Rat | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 10333 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction