missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse TLR4 Antibody, R&D Systems™

Rabbit Monoclonal Antibody
Brand: R&D Systems MAB27591-SP
This item is not returnable.
View return policy
Description
TLR4 Monoclonal specifically detects TLR4 in Mouse samples. It is validated for Flow Cytometry, Immunocytochemistry.
Specifications
| TLR4 | |
| Monoclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with Sodium Azide | |
| ARMD10, CD284, CD284 antigen, hTollhomolog of Drosophila toll, TOLL, toll-like receptor 4 | |
| Rabbit | |
| Protein A or G purified from cell culture supernatant | |
| RUO | |
| 7099 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG |
| Flow Cytometry, Immunocytochemistry | |
| 1203B | |
| Flow Cytometry 0.25 ug/10^6 cells, Immunocytochemistry 1-25 ug/mL | |
| Q9QUK6 | |
| Tlr4 | |
| Mouse myeloma cell line NS0-derived recombinant mouse TLR4 Asn26-Lys629 Accession # Q9QUK6 | |
| 25 μg | |
| Primary | |
| Detects mouse TLR4 in direct ELISAs. | |
| Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction