missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse TLR2 Antibody, R&D Systems™

Rat Monoclonal Antibody has been used in 5 publications
Brand: R&D Systems MAB1530
This item is not returnable.
View return policy
Description
TLR2 Monoclonal specifically detects TLR2 in Mouse samples. It is validated for Flow Cytometry, CyTOF-ready.
Specifications
| TLR2 | |
| Monoclonal | |
| LYOPH | |
| Flow Cytometry 0.25 ug/10^6 cells, CyTOF-ready | |
| Q9QUN7 | |
| TLR2 | |
| Mouse myeloma cell line NS0-derived recombinant mouse TLR2 Gln25-Leu590 Accession # Q9QUN7 | |
| 500 μg | |
| Primary | |
| Detects mouse TLR2 in direct ELISAs. In direct ELISAs, this antibody does not cross-react with recombinant mouse (rm) TLR1, recombinant human (rh) TLR2, rhTLR3, rhTLR4, or rmTLR6. | |
| Mouse | |
| Purified |
| Flow Cytometry, CyTOF | |
| 203325 | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| CD282, CD282 antigen, TIL4CD282, toll/interleukin 1 receptor-like 4, Toll/interleukin-1 receptor-like protein 4, toll-like receptor 2 | |
| Rat | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 7097 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction