missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Mouse TIMP-2 DuoSet ELISA

Product Code. 30000474
Click to view available options
Quantity:
1 Kit (for 5 Plates)
Unit Size:
Pack of 5
This item is not returnable. View return policy

Product Code. 30000474

Brand: R&D Systems™ DY630405

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Includes

Capture antibody, detection antibody, recombinant standard, streptavidin conjugated to horseradish-peroxidase (Streptavidin-HRP) and development protocols

Mouse TIMP-2 DuoSet ELISA from the most referenced ELISA manufacturer. Highly validated for accurate quantitation and long-term reproducibility.

The Mouse TIMP-2 DuoSet ELISA is a Solid Phase Sandwich ELISA that quantifies mouse TIMP-2.
TRUSTED_SUSTAINABILITY

Specifications

Quantity 1 Kit (for 5 Plates)
Species Mouse
Conjugate Biotin
Binding Target TIMP-2
For Use With (Application) ELISA
Gene ID (Entrez) 7077
Synonym CSC-21Ktissue inhibitor of metalloproteinase 2, metalloproteinase inhibitor 2, TIMP metallopeptidase inhibitor 2, TIMP2, TIMP-2, Tissue inhibitor of metalloproteinases 2
Content And Storage Store the unopened product at 2°C to 8°C. Do not use past expiration date.
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.