Learn More
ACROBiosystems Mouse TIGIT Protein, Fc Tag
Description
T-cell immunoreceptor with Ig and ITIM domains (TIGIT) is also known as V-set and immunoglobulin domain-containing protein 9 (VSIG9), V-set and transmembrane domain-containing protein 3 (VSTM3),which belongs to single-pass type I membrane protein containing an immunoglobulin variable domain, a transmembrane domain and an immunoreceptor tyrosine-based inhibitory motif (ITIM). TIGIT is expressed at low levels on peripheral memory and regulatory CD4+ T-cells and NK cells and is up-regulated following activation of these cells (at protein level). TIGIT binds with high affinity to the poliovirus receptor (PVR) which causes increased secretion of IL10 and decreased secretion of IL12B and suppresses T-cell activation by promoting the generation of mature immunoregulatory dendritic cells.
Specifications
Specifications
| Accession Number | NP_001139797.1 |
| Molecular Weight (g/mol) | 39.5 kDa |
| Quantity | 1 mg |
| Source | Mouse TIGIT, Fc Tag (TIT-M5257) is expressed from human 293 cells (HEK293). It contains AA Gly 26 - Thr 143 (Accession # NP_001139797.1). |
| Immunogen | Gly 26 - Thr 143 |
| Storage Requirements | -20°C |
| Gene Alias | TIGIT,VSIG9,VSTM3 |
| Common Name | TIGIT |
| Conjugate | Unconjugated |
| Species | Mouse |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.