missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Mouse Tie-2 Quantikine ELISA Kit

Product Code. 18782264
Click to view available options
Quantity:
1 x 96-well Plate
Unit Size:
1 set
This item is not returnable. View return policy

Product Code. 18782264

Brand: R&D Systems™ MTE200

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Tie-2 Quantikine ELISA Kit from the most referenced ELISA manufacturer. Highly validated for accurate quantitation and long-term reproducibility.

The Mouse Tie-2 Quantikine ELISA Kit is a Solid Phase Sandwich ELISA that quantifies mouse Tie-2 in cell culture supernates (50 ul), tissue homogenates (50 ul), serum (25 ul), edta plasma (25 ul), heparin plasma (25 ul).
TRUSTED_SUSTAINABILITY

Specifications

Assay Range 125.00 - 8000pg/mL
Assay Sensitivity 40.2pg/mL
Conjugate HRP
Sample Type Cell Culture Supernatant, Plasma (EDTA, Heparin), Serum, Tissue Homogenates
Specificity Natural and Recombinant Mouse Tie-2
For Use With (Application) ELISA
Gene ID (Entrez) 21687
Quantity 1 x 96-well Plate
Sample Volume Cell Culture Supernates (50uL),Tissue Homogenates (50uL),Serum (25uL),EDTA Plasma (25uL),Heparin Plasma (25uL)
Storage Requirements Store the unopened product at 2° to 8°C. Do not use past expiration date.
Synonym angiopoietin-1 receptor, CD202B, CD202b antigen, EC 2.7.10, EC 2.7.10.1, hTIE2, p140 TEK, soluble TIE2 variant 1, soluble TIE2 variant 2, TEK, TEK tyrosine kinase, endothelial, Tie2, TIE-2, TIE2CD202b, Tunica interna endothelial cell kinase, Tyrosine-protein kinase receptor TEK, Tyrosine-protein kinase receptor TIE-2, venous malformations, multiple cutaneous and mucosal, VMCM, VMCM1
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.