missing translation for 'onlineSavingsMsg'
Learn More

Proteintech Mouse Thrombopoietin ELISA Kit

Product Code. 17254886 Shop All Proteintech Products
Change view
Click to view available options
Gene ID (Entrez):
22018
Unit Size:
96 reactions
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Gene ID (Entrez) Unit Size
17254886 22018 96 reactions
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17254886 Supplier Proteintech Supplier No. KE1005996T
 Restricted Item
Estimated Shipment: 26-10-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Thrombopoietin (TPO) is produced in the liver by both parenchymal cells and sinusoidal endothelial cells, as well as in the kidney by proximal convoluted tubule cells.

It is a primary regulator of megakaryocyte development and platelet production in mammals. TPO binds the TPO receptor, activates JAK and STAT pathways, thus stimulating megakaryocyte growth and platelet production. Additionally, as TPO is vital for the maintenance of haematopoietic stem cells, it can truly be described as a pan- haematopoietic cytokine.

Specifications

Gene ID (Entrez) 22018
compliance-icons compliance-icons
Product Identifier
  • 96RXN Mouse Thrombopoietin ELISA Kit
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H302-Harmful if swallowed.
  • H315-Causes skin irritation.
  • H318-Causes serious eye damage.
  • H319-Causes serious eye irritation.
  • H332-Harmful if inhaled.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P201-Obtain special instructions before use.
  • P202-Do not handle until all safety precautions have been read and understood.
  • P234-Keep only in original container.
  • P264-Wash skin thoroughly after handling.
  • P270-Do not eat, drink or smoke when using this product.
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
  • P330-Rinse mouth.
  • P362-Take off contaminated clothing.
  • P405-Store locked up.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contain: Sulphuric Acid, Tetramethylbenzidine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.