missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse Thrombomodulin/BDCA-3 Alexa Fluor 405-conjugated Antibody, R&D Systems™

Rat Monoclonal Antibody
Brand: R&D Systems FAB3894V-100UG
This item is not returnable.
View return policy
Description
Thrombomodulin/BDCA-3 Monoclonal specifically detects Thrombomodulin/BDCA-3 in Mouse samples. It is validated for Flow Cytometry.
Specifications
| Thrombomodulin/BDCA-3 | |
| Monoclonal | |
| 0.2 mg/mL | |
| Flow Cytometry 0.25-1 ug/10^6 cells | |
| P15306 | |
| THBD | |
| Mouse myeloma cell line NS0-derived recombinant mouse Thrombomodulin/BDCA-3 Leu17-Ser517 Accession # P15306 | |
| RUO | |
| 7056 | |
| Mouse | |
| IgG2b |
| Flow Cytometry | |
| 461714 | |
| Alexa Fluor 405 | |
| Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. with 0.09% Sodium Azide | |
| BDCA3, BDCA-3, CD141, CD141 antigen, fetomodulin, THBD, THRM, thrombomodulin, TM | |
| Rat | |
| 100 μg | |
| Primary | |
| Detects mouse Thrombomodulin/BDCA-3 in direct ELISAs and Western blots. In direct ELISAs and Western blots, no cross-reactivity with recombinant human (rh) Thrombomodulin is observed. | |
| Store the unopened product at 2 - 8 degreesC. Do not use past expiration date. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only (RUO)
Spot an opportunity for improvement?Share a Content Correction