missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ Mouse Thrombin-antithrombin (TAT) complexes ELISA Kit (Chemiluminescence)
Shop All Bio Techne ProductsSpecifications
Specifications
| Assay Range | 7.81 to 500 pg/mL |
| Assay Sensitivity | 4.69 pg/mL |
| Product Type | ELISA Kit (Chemiluminescence) |
| Sample Type | Serum, Plasma, Biological Fluids |
| Specificity | This kit recognizes Mouse TAT in samples. No significant cross-reactivity or interference between Mouse TAT and analogues was observed. |
| For Use With (Application) | ELISA |
| Quantity | 1 Kit |
| Research Discipline | Apoptosis, Breast Cancer, Cancer, Signal Transduction |
| Storage Requirements | Storage of components varies. See protocol for specific instructions. |
| Target | Mouse |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?