missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ eBioscience™ Mouse Th17 Cytokine Staining Panel

Product Code. 15561227
Click to view available options
:
Mouse Th17 Cytokine Staining Panel
This item is not returnable. View return policy

Product Code. 15561227

Brand: Invitrogen™ 88841144

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Includes

  • Anti-Mouse IL-17A (eBio17B7) FITC : 25 ug ([optimal] = 0.25 μg)*
  • Anti-Mouse IL-17F (eBio18F10) PE : 25 ug ([optimal] = 0.5 μg)*
  • Anti-Mouse IL-22 (1H8PWSR) PerCP-eFluor 710 : 25 ug ([optimal] = 0.125 μg)*
  • Anti-Mouse IL-21 (FFA21) APC : 25 ug ([optimal] = 0.125 μg)*
  • Anti-Mouse CD4 (RM4-5) eFluor 450 : 25 ug ([optimal] = 0.125 μg)*
  • IC Fixation Buffer
  • Permeabilization Buffer (10X)
  • *The optimal concentration is based on 1 million cells in a 100 μL total staining volume.

Includes all reagents needed for simultaneous flow cytometric detection of all the major cytokines produced by the Th17 lineage (IL-17A, IL-17F, IL-21 and IL-22).

Also included: An anti-CD4 antibody that can be used after fixation and permeabilization, as well as IC Fixation and Permeabilization Buffers.
TRUSTED_SUSTAINABILITY

Specifications

Detects IL-17A, IL-17F, IL-21, IL-22
For Use With (Application) Intracellular Staining Followed by Flow Cytometric Analysis
Storage Buffer Aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
Content And Storage 2°C to 8°C. Do not freeze. Light sensitive material.

For Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.