Learn More
Invitrogen™ TGF beta-1 Mouse ELISA Kit
For the quantitative detection of mouse TGF-beta1.
£451.00 - £2400.00
Specifications
Accession Number | P04202 |
---|---|
Assay Range | 31.25-2,000 pg/mL |
Assay Sensitivity | 7.8 pg/mL |
Product Type | ELISA Kit |
Sample Type | Plasma, Serum, Supernatant |
Includes: Aluminium pouch(es) with a Microwell Plate coated with monoclonal antibody to mouse TGF-beta1
Biotin-Conjugate anti-mouse TGF-beta1 polyclonal antibody
Streptavidin-HRP
Mouse TGF-beta1 Standard lyophilized, 4ng/mL upon reconstitution
Assay Buffer Concentrate 20x (PBS with 1% Tween 20 and 10% BSA)
Wash Buffer Concentrate 20x (PBS with 1% Tween 20)
]Substrate Solution (tetramethyl-benzidine)
Stop Solution (1M Phosphoric acid)
Adhesive Films
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15512027
|
Invitrogen™
BMS608-4 |
96 Tests |
£451.00
96 tests |
Estimated Shipment: 21-05-2024
Log in to see stock. |
|||||
15542027
|
Invitrogen™
BMS608-4TWO |
2 x 96 Tests |
£580.00
Pack of 2 |
In Stock
Log in to see stock. |
|||||
15522027
|
Invitrogen™
BMS608-4FIVE |
5 x 96 Tests |
£1444.00
Pack of 5 |
In Stock
Log in to see stock. |
|||||
15532027
|
Invitrogen™
BMS608-4TEN |
10 x 96 Tests |
£2400.00
Pack of 10 |
Estimated Shipment: 04-06-2024
Log in to see stock. |
|||||
Description
The Mouse Transforming Growth Factor beta 1 (Ms TGFβ1) ELISA quantitates Ms TGFβ1in Mouse serum, plasma, buffered solution, or cell culture medium. The assay will exclusively recognize both natural and recombinant Mouse TGFβ1. Principle of the method The Mouse TGFβ1 solid-phase sandwich ELISA (enzyme-linked immunosorbent assay) is designed to measure the amount of the target bound between a matched antibody pair. A target-specific antibody has been pre-coated in the wells of the supplied microplate. Samples, standards, or controls are then added into these wells and bind to the immobilized (capture) antibody. The sandwich is formed by the addition of the second (detector) antibody, a substrate solution is added that reacts with the enzyme-antibody-target complex to produce measurable signal. The intensity of this signal is directly proportional to the concentration of target present in the original specimen. Rigorous validation Each manufactured lot of this ELISA kit is quality tested for criteria such as sensitivity, specificity, precision, and lot-to-lot consistency. See manual for more information on validation.
Platinum Kits undergo stringent in-house development, manufacture and quality-validation processes ensuring highest possible product performance and optimal lot-to-lot consistencySpecifications
P04202 | |
7.8 pg/mL | |
Plasma, Serum, Supernatant | |
ELISA | |
21803 | |
7.9% | |
HRP | |
Plasma, 20 μL; Serum, 20 μL; Supernatant, 20 μL | |
Camurati-Engelmann disease, DPD1 | |
1 hr. 20 min. |
31.25-2,000 pg/mL | |
ELISA Kit | |
Mouse | |
Colorimetric Microplate Reader | |
5.8% | |
Pre-coated 96 well plate, Standard, Sample Diluent, Assay Buffer concentrate, HRP-conjugated Detection Antibody, Wash Buffer, Chromogen, Stop Solution, Adhesive Plate Covers | |
RUO | |
2°C to 8°C | |
Mouse | |
4 hr. |