missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Mouse TFPI DuoSet ELISA

Product Code. 30021277
Click to view available options
Quantity:
15 x 96-well Plates
Unit Size:
1 set
This item is not returnable. View return policy

Product Code. 30021277

Brand: R&D Systems™ DY2975

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Sufficient for at least 15 96-well plates

The Mouse TFPI DuoSet ELISA is a Solid Phase Sandwich ELISA that quantifies mouse TFPI.
TRUSTED_SUSTAINABILITY

Specifications

Quantity 15 x 96-well Plates
Conjugate Biotin
Assay Range 62.50 to 4000 pg/mL
For Use With (Application) ELISA
Gene ID (Entrez) 21788
Content And Storage Store the unopened product at 2°C to 8°C. Do not use past expiration date.
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.