missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse TDRD1 Antibody, R&D Systems™
Sheep Polyclonal Antibody
Brand: R&D Systems AF7320
This item is not returnable.
View return policy
Description
TDRD1 Polyclonal specifically detects TDRD1 in Mouse samples. It is validated for Western Blot.
Specifications
| TDRD1 | |
| Polyclonal | |
| Unconjugated | |
| Q99MV1 | |
| TDRD1 | |
| E. coli-derived recombinant mouse TDRD1 Asn831-Lys960 Accession # Q99MV1 | |
| 100 μg | |
| Primary | |
| Detects mouse TDRD1 in direct ELISAs and Western blots. In direct ELISAs, approximately 60% cross-reactivity with recombinant human TDRD1 is observed. | |
| Mouse | |
| IgG |
| Western Blot | |
| LYOPH | |
| Western Blot 1 ug/mL | |
| CT41.1, CT41.1Cancer/testis antigen 41.1, FLJ21082, tudor domain containing 1, tudor domain containing protein 1, tudor domain-containing protein 1 | |
| Sheep | |
| Affinity Purified | |
| RUO | |
| 56165 | |
| Sterile PBS to a final concentration of 0.2 mg/mL. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction