missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse TCAM-1 Antibody, R&D Systems™
Goat Polyclonal Antibody
Brand: R&D Systems AF3546
This item is not returnable.
View return policy
Description
TCAM-1 Polyclonal specifically detects TCAM-1 in Mouse samples. It is validated for Western Blot.
Specifications
| TCAM-1 | |
| Polyclonal | |
| Unconjugated | |
| AAH50746 | |
| Goat | |
| Affinity Purified | |
| RUO | |
| 146771 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
| Western Blot | |
| LYOPH | |
| Western Blot 0.1 ug/mL | |
| Tcam1 | |
| Mouse myeloma cell line NS0-derived recombinant mouse TCAM-1 Ala18-Gly481 Accession # AAH50746 | |
| 100 μg | |
| Primary | |
| Detects mouse TCAM-1 in direct ELISAs and Western blots. | |
| Mouse | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction