missing translation for 'onlineSavingsMsg'
Learn More

Mouse T-bet/TBX21 Antibody, R&D Systems™

Rat Monoclonal Antibody

£206.00 - £476.00

Specifications

Antigen T-bet/TBX21
Clone 1087415
Dilution Western Blot 2 μg/mL, Immunohistochemistry 3-25 μg/mL
Applications Western Blot, Immunohistochemistry
Classification Monoclonal
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
30279867
View Documents Promotion Details
R&D Systems
MAB11627-SP
25 μg
£206.00
25µg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
30279868
View Documents
R&D Systems
MAB11627-100
100 μg
£476.00
100µg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

T-bet/TBX21 Monoclonal antibody specifically detects T-bet/TBX21 in Mouse samples. It is validated for Western Blot,Immunohistochemistry
Specifications

Specifications

T-bet/TBX21
Western Blot 2 μg/mL, Immunohistochemistry 3-25 μg/mL
Monoclonal
Purified
RUO
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose.
Tbet, T-bet, TBETT-PET, TBLYM, TBLYMT-cell-specific T-box transcription factor T-bet, T-box 21, T-box expressed in T cells, T-box protein 21, T-box transcription factor TBX21, TBX21, T-PET, Transcription factor TBLYM
Synthetic Peptide, Accession # Q9JKD8
Primary
Reconstitute lyophilized material at 0.2 mg/mL in sterile PBS. For liquid material, refer to CoA for concentration.
1087415
Western Blot, Immunohistochemistry
Unconjugated
Rat
Mouse
Q9JKD8
30009
IgG2b
Protein A or G purified from cell culture supernatant
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20°C to -70 °C as supplied. 1 month, 2 to 8 °C under sterile conditions after reconstitution. 6 months, -20°C to -70 °C under sterile conditions after reconstitution.
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.