missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse Syndecan-3 Antibody, R&D Systems™

Rat Monoclonal Antibody has been used in 3 publications
Brand: R&D Systems MAB2734-SP
This item is not returnable.
View return policy
Description
SDC3 Monoclonal specifically detects SDC3 in Mouse samples. It is validated for Western Blot.
Specifications
| SDC3 | |
| Monoclonal | |
| LYOPH | |
| Western Blot 1 ug/mL | |
| Q64519 | |
| SDC3 | |
| Mouse myeloma cell line NS0-derived recombinant mouse Syndecan-3 Ala45-Glu384 Accession # Q64519 | |
| 25 μg | |
| Primary | |
| Detects mouse Syndecan-3 in direct ELISAs and Western blots. In direct ELISAs and Western blots, no cross-reactivity with recombinant mouse (rm) Syndecan-1, rmSyndecan-4, or recombinant human Syndecan-3 is observed. | |
| Mouse | |
| Purified |
| Western Blot | |
| 312607 | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| KIAA0468, N-syndecan, SDC3, SDCN, SYND3syndecan proteoglycan 3, syndecan 3, syndecan 3 (N-syndecan), syndecan neural type, syndecan-3, Syndecan3 | |
| Rat | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 9672 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction