missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Mouse Syndecan-2/CD362 (NP_032330) VersaClone cDNA
Click to view available options
Quantity:
10 μg
Unit Size:
10µg
Description
- Also known as: Hspg1; Synd2; AA960457; syndecan-2; 4833414L08Rik
- SDC2 is a type I transmembrane heparan sulfate proteoglycan and is a member of the syndecan proteoglycan family.
- The syndecans mediate cell binding, cell signaling, and cytoskeletal organization and syndecan receptors are required for internalization of the HIV-1 tat protein.
- SDC2 functions as an integral membrane protein and participates in cell proliferation, cell migration and cell-matrix interactions via its receptor for extracellular matrix proteins.
- Altered SDC2 expression has been detected in several different tumor types.
- The insert is sequence verified and the entire plasmid DNA and insert translated sequences are provided.
- For reasearch use only. Not for use in humans.
Specifications
Specifications
| Gene | mSdc2 |
| Accession Number | NP_032330 |
| Concentration | 10 μg at 0.2 μg/μL |
| Formulation | cDNA is provided in 10 mM Tris-Cl, pH 8.5 |
| Insert Type | 622 bp |
| Gene ID (Entrez) | 6383 |
| Content And Storage | Store the unopened product at -20°C to -70°C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date. |
| Species | Mouse |
| Quantity | 10 μg |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction