missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse Syndecan-1/CD138 Antibody, R&D Systems™

Rat Monoclonal Antibody
£170.00 - £368.00
Specifications
| Antigen | Syndecan-1/CD138 |
|---|---|
| Clone | 320611 |
| Concentration | LYOPH |
| Dilution | Western Blot 1 ug/mL, Immunohistochemistry 1-25 ug/mL, Immunoprecipitation 25 ug/mL |
| Applications | Western Blot, Immunohistochemistry, Immunoprecipitation |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18781879
![]() |
R&D Systems
MAB29661-SP |
25 μg |
£170.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18132351
|
R&D Systems
MAB29661 |
100 μg |
£368.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Syndecan-1/CD138 Monoclonal specifically detects Syndecan-1/CD138 in Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunoprecipitation.Specifications
| Syndecan-1/CD138 | |
| LYOPH | |
| Western Blot, Immunohistochemistry, Immunoprecipitation | |
| Unconjugated | |
| Rat | |
| Mouse | |
| P18828 | |
| 6382 | |
| Mouse myeloma cell line NS0-derived recombinant mouse Syndecan-1/CD138 isoform 1 Gln18-Glu252 (predicted) Accession # P18828 | |
| Primary | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Detects mouse Syndecan-1/CD138 in direct ELISAs and Western blots. In direct ELISAs and Western blots, no cross-reactivity with recombinant human Syndecan-1, recombinant mouse (rm) Syndecan-2, rmSyndecan-3, or rmSyndecan-4 is observed. |
| 320611 | |
| Western Blot 1 ug/mL, Immunohistochemistry 1-25 ug/mL, Immunoprecipitation 25 ug/mL | |
| Monoclonal | |
| Purified | |
| RUO | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| CD138, CD138 antigen, SDC, SDC1, SYND1heparan sulfate proteoglycan fibroblast growth factor receptor, syndecan, syndecan 1, syndecan proteoglycan 1, syndecan-1, Syndecan1 | |
| SDC1 | |
| IgG1 | |
| Protein A or G purified from hybridoma culture supernatant | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title