missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse Sulfatase Modifying Factor 1/SUMF1 Antibody, R&D Systems™
Goat Polyclonal Antibody
Brand: R&D Systems AF3335
Additional Details : Weight : 0.00970kg
Description
Sulfatase Modifying Factor 1/SUMF1 Polyclonal specifically detects Sulfatase Modifying Factor 1/SUMF1 in Mouse samples. It is validated for Western Blot.Specifications
Sulfatase Modifying Factor 1/SUMF1 | |
Polyclonal | |
Unconjugated | |
Q8R0F3 | |
SUMF1 | |
Mouse myeloma cell line NS0-derived recombinant mouse Sulfatase Modifying Factor 1/SUMF1 Ser32-Asp372 Accession # Q8R0F3 | |
100 μg | |
Primary | |
Detects mouse Sulfatase Modifying Factor 1/SUMF1 in direct ELISAs and Western blots. In direct ELISAs and Western blots, approximately 50% cross-reactivity with recombinant human SUMF1 is observed and 10% cross-reactivity with recombinant mouse SUMF2 is observed. | |
Mouse | |
IgG |
Western Blot | |
LYOPH | |
Western Blot 0.1 ug/mL | |
AAPA3037, EC 1.8.99.-, FGE1, FGEC-alpha-formylglycine-generating enzyme 1, FGly-generating enzyme, MGC131853, MGC150436, sulfatase modifying factor 1, sulfatase-modifying factor 1, UNQ3037 | |
Goat | |
Affinity Purified | |
RUO | |
285362 | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
For Research Use Only