missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ Mouse Stomach Whole Tissue Lysate (Adult Whole Normal)

Product Code. 18640331 Shop All Bio Techne Products
Click to view available options
Quantity:
1 mg
Unit Size:
1mg
This item is not returnable. View return policy

Product Code. 18640331

Brand: Novus Biologicals™ NB82059671

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Specifications

Preparation Method Western Blot Electrophoresis
Quantity 1 mg
Content And Storage Store at -80°C. Avoid freeze/thaw cycles.
Target Stomach
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.