missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Mouse ST2/IL-33R Quantikine ELISA Kit

Product Code. 18772064
Change view
Click to view available options
Quantity:
1 x 96-well Plate
Unit Size:
1 set
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18772064 1 x 96-well Plate 1 set
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18772064 Supplier R&D Systems™ Supplier No. MST200

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse ST2/IL-33 R Quantikine ELISA Kit from the most referenced ELISA manufacturer. Highly validated for accurate quantitation and long-term reproducibility.

The Mouse ST2/IL-33 R Quantikine ELISA Kit is a Solid Phase Sandwich ELISA that quantifies mouse ST2/IL-33R in cell culture supernates (50 ul), serum (10 ul), edta plasma (10 ul), heparin plasma (10 ul).
TRUSTED_SUSTAINABILITY

Specifications

Assay Range 0.16 - 10ng/mL
Assay Sensitivity 0.059ng/mL
Conjugate HRP
Sample Type Cell Culture Supernatant, Plasma (EDTA), Plasma (Heparin), Serum
Specificity Natural and Recombinant Mouse ST2
For Use With (Application) ELISA
Gene ID (Entrez) 17082
Quantity 1 x 96-well Plate
Sample Volume Cell Culture Supernates (50uL),Serum (10uL),EDTA Plasma (10uL),Heparin Plasma (10uL)
Storage Requirements Store the unopened product at 2° to 8°C. Do not use past expiration date.
Synonym DER4, DER4ST2growth stimulation-expressed, FIT-1, IL-1 R4, IL1R4, IL-1R4, IL-1 RL1, IL-33 R, IL33R, IL-33 R, interleukin 1 receptor-like 1, interleukin 1 receptor-related protein, interleukin-1 receptor-like 1, Ly84, MGC32623, Protein ST2, ST2L, ST2V, T1, T1homolog of mouse growth stimulation-expressed
Target ST2/IL-33R
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.