missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Mouse ST2/IL-33R DuoSet ELISA
Click to view available options
Conjugate:
Biotin
Unit Size:
1 set
Description
The Mouse ST2/IL-33 R DuoSet ELISA is a Solid Phase Sandwich ELISA that quantifies mouse ST2/IL-33R.
Specifications
Specifications
| Conjugate | Biotin |
| Assay Range | 0.31 to 20 ng/mL |
| For Use With (Application) | ELISA |
| Gene ID (Entrez) | 17082 |
| Synonym | DER4, DER4ST2growth stimulation-expressed, FIT-1, IL-1 R4, IL1R4, IL-1R4, IL-1RL1, IL-33 R, IL33R, IL-33R, interleukin 1 receptor-like 1, interleukin 1 receptor-related protein, interleukin-1 receptor-like 1, Ly84, MGC32623, Protein ST2, ST2L, ST2V, T1, T1homolog of mouse growth stimulation-expressed |
| Content And Storage | Store the unopened product at 2°C to 8°C. Do not use past expiration date. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction