missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse SorCS3 Antibody, R&D Systems™

Rat Monoclonal Antibody
Brand: R&D Systems MAB30671-SP
This item is not returnable.
View return policy
Description
SorCS3 Monoclonal specifically detects SorCS3 in Mouse samples. It is validated for Western Blot, Immunohistochemistry.
Specifications
| SorCS3 | |
| Monoclonal | |
| LYOPH | |
| Western Blot 1 ug/mL, Immunohistochemistry 8-25 ug/mL | |
| Q8VI51 | |
| SORCS3 | |
| Mouse myeloma cell line NS0-derived recombinant mouse SorCS3 Glu34-Ser1122 Accession # Q8VI51 | |
| 25 μg | |
| Primary | |
| Detects human and mouse SorCS3 in direct ELISAs and Western blots. In direct ELISAs and Western blots, no cross-reactivity with recombinant human SorCS1 is observed. | |
| Mouse | |
| Purified |
| Western Blot, Immunohistochemistry | |
| 339615 | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| 6330404A12Rik, KIAA1059SORCS, sortilin-related VPS10 domain containing receptor 3, VPS10 domain receptor protein SORCS 3 (SORCS3), VPS10 domain-containing receptor SorCS3 | |
| Rat | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 22986 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction