missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse Soggy-1/DkkL1 Biotinylated Antibody, R&D Systems™
Goat Polyclonal Antibody
Brand: R&D Systems BAF1508
This item is not returnable.
View return policy
Description
Soggy-1/DkkL1 Polyclonal specifically detects Soggy-1/DkkL1 in Mouse samples. It is validated for Western Blot.
Specifications
| Soggy-1/DkkL1 | |
| Polyclonal | |
| Biotin | |
| AAF02679 | |
| DKKL1 | |
| Mouse myeloma cell line NS0-derived recombinant mouse Soggy-1/DkkL1 Leu21-Leu230 Accession # AAF02679 | |
| 50 μg | |
| Primary | |
| Detects mouse Soggy-1/DkkL1 in Western blots. In Western blots, approximately 5% cross-reactivity with recombinant human Soggy-1 is observed. | |
| Mouse | |
| IgG |
| Western Blot | |
| LYOPH | |
| Western Blot 0.1 ug/mL | |
| Cancer/testis antigen 34, CT34dickkopf-like 1 (soggy), dickkopf-like 1, DkkL1, Protein soggy-1, SGY, SGY1, SGY-1dickkopf-like protein 1, SGY1soggy, soggy-1, Soggy1 | |
| Goat | |
| Affinity Purified | |
| RUO | |
| 27120 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction