Learn More
Mouse SOD3/EC-SOD Antibody, R&D Systems™
GREENER_CHOICE
Goat Polyclonal Antibody has been used in 4 publications
Brand:  R&D Systems AF4817-SP
Product Code. 18725157
  • £167.00 / 25µg
Description
SOD3/EC-SOD Polyclonal specifically detects SOD3/EC-SOD in Mouse samples. It is validated for Western Blot.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
SOD3/EC-SOD
Polyclonal
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
ECSOD, EC-SOD, EC-SODEC 1.15.1.1, extracellular superoxide dismutase [Cu-Zn], MGC20077, superoxide dismutase 3, extracellular
Goat
Affinity Purified
RUO
6649
Reconstitute at 0.2 mg/mL in sterile PBS.
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Western Blot
LYOPH
Western Blot 1 ug/mL
O09164
SOD3
E. coli-derived recombinant mouse SOD3/EC-SOD Ser25-Thr251 Accession # O09164
25 μg
Primary
Detects mouse SOD3/EC-SOD in Western blots. In Western blots, less than 1% cross-reactivity with recombinant mouse SOD1 or SOD2 is observed.