missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse SLPI Antibody, R&D Systems™

Goat Polyclonal Antibody has been used in 2 publications
Brand: R&D Systems AF1735
This item is not returnable.
View return policy
Description
SLPI Polyclonal specifically detects SLPI in Mouse samples. It is validated for Western Blot.
Specifications
| SLPI | |
| Polyclonal | |
| Unconjugated | |
| ALK1, ALPProtease inhibitor WAP4, antileukoproteinase, BLPISeminal proteinase inhibitor, HUSI, HUSI-I, Mucus proteinase inhibitor, secretory leukocyte peptidase inhibitor, Secretory leukocyte protease inhibitor, secretory leukocyte protease inhibitor (antileukoproteinase), WAP four-disulfide core domain protein 4, WAP4HUSI-1, WFDC4MPI | |
| Goat | |
| Affinity Purified | |
| RUO | |
| 6590 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
| Western Blot | |
| LYOPH | |
| Western Blot 0.1 ug/mL | |
| SLPI | |
| Mouse myeloma cell line NS0-derived mouse SLPI | |
| 100 μg | |
| Primary | |
| Detects mouse SLPI in direct ELISAs and Western blots. In these formats, less than 5% cross-reactivity with recombinant human SLPI is observed. | |
| Mouse | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction