missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse Slit3 Antibody, R&D Systems™

Rat Monoclonal Antibody
Brand: R&D Systems MAB3629
This item is not returnable.
View return policy
Description
Slit3 Monoclonal specifically detects Slit3 in Mouse samples. It is validated for Immunohistochemistry.
Specifications
| Slit3 | |
| Monoclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| KIAA0814, MEGF5, MEGF5Multiple epidermal growth factor-like domains protein 5, Multiple EGF-like domains protein 5, SLIL2, SLIL2FLJ10764, slit homolog 3 (Drosophila), slit homolog 3 protein, SLIT1, slit2, Slit-3slit (Drosophila) homolog 3 | |
| Rat | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 6586 | |
| Sterile PBS to a final concentration of 0.5 mg/mL. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG2b |
| Immunohistochemistry | |
| 782619 | |
| Immunohistochemistry 8-25 ug/mL | |
| Q9WVB4 | |
| SLIT3 | |
| Mouse myeloma cell line NS0-derived recombinant mouse Slit3 Ser27-His901 Accession # Q9WVB4 | |
| 100 μg | |
| Primary | |
| Detects mouse Slit3 in direct ELISAs. In direct ELISAs, no cross-reactivity with recombinant mouse (rm) Slit1, rmSlit2, or recombinant human Slit3 is observed. | |
| Mouse | |
| Supernatant |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction