missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse SIGNR1/CD209b Antibody, R&D Systems™

Goat Polyclonal Antibody has been used in 3 publications
Brand: R&D Systems AF1836
This item is not returnable.
View return policy
Description
SIGNR1/CD209b Polyclonal specifically detects SIGNR1/CD209b in Mouse samples. It is validated for Western Blot, Adhesion Blockade.
Specifications
| SIGNR1/CD209b | |
| Polyclonal | |
| Unconjugated | |
| Q8CJ91 | |
| Cd209b | |
| Mouse myeloma cell line NS0-derived recombinant mouse SIGNR1 Ser76-Gly325 Accession # Q8CJ91 | |
| 100 μg | |
| Primary | |
| Detects mouse SIGNR1 in direct ELISAs and Western blots. In direct ELISAs, approximately 20% cross-reactivity with recombinant mouse (rm) SIGNR3 and rmSIGNR4 is observed, and approximately 10% cross-reactivity with recombinant human DC-SIGNR and rmDC-SIGN is observed. | |
| Mouse | |
| IgG |
| Western Blot, Adhesion Blockade | |
| LYOPH | |
| Western Blot 0.1 ug/mL, Adhesion Blockade 10 ug/mL | |
| CD209 antigen like protein B, CD209b, DC SIGN related protein 1, DC SIGNR1, OtB7 | |
| Goat | |
| Affinity Purified | |
| RUO | |
| 170786 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction