missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse SIGNR1/CD209b Antibody, R&D Systems™
Rat Monoclonal Antibody has been used in 3 publications
Brand: R&D Systems MAB1836
This item is not returnable.
View return policy
Description
SIGNR1/CD209b Monoclonal specifically detects SIGNR1/CD209b in Mouse samples. It is validated for Western Blot.
Specifications
| SIGNR1/CD209b | |
| Monoclonal | |
| LYOPH | |
| Western Blot 1 ug/mL | |
| Q8CJ91 | |
| Cd209b | |
| Mouse myeloma cell line NS0-derived recombinant mouse SIGNR1/CD209b Ser76-Gly325 Accession # Q8CJ91 | |
| 100 μg | |
| Primary | |
| Detects mouse SIGNR1/CD209b in direct ELISAs and Western blots. In direct ELISAs and Western blots, no cross-reactivity with recombinant mouse SIGNR4 is observed. | |
| Mouse | |
| Purified |
| Western Blot | |
| 258829 | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| CD209 antigen like protein B, CD209b, DC SIGN related protein 1, DC SIGNR1, OtB7 | |
| Rat | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 170786 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction