missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse Siglec-F Antibody, R&D Systems™
Goat Polyclonal Antibody has been used in 1 publication
Brand: R&D Systems AF1706
Additional Details : Weight : 0.00970kg
Description
Siglec-F Polyclonal specifically detects Siglec-F in Mouse samples. It is validated for Western Blot.Specifications
Siglec-F | |
Polyclonal | |
Unconjugated | |
Q920G3 | |
Siglecf | |
Mouse myeloma cell line NS0-derived recombinant mouse Siglec-F Asp18-Thr437 Accession # Q920G3 | |
100 μg | |
Primary | |
Detects mouse Siglec-F in direct ELISAs and Western blots. In direct ELISAs and Western blots, less than 1% cross-reactivity with recombinant mouse (rm) Siglec-2, rmSiglec-3, recombinant human (rh) Siglec-5, rhSiglec-7, rhSiglec-9, and rhSiglec-10 is observed. | |
Mouse | |
IgG |
Western Blot | |
LYOPH | |
Western Blot 0.1 ug/mL | |
SAF2, sialic acid binding Ig like lectin 8, SIGLEC-8, SIGLEC8L, SiglecF | |
Goat | |
Affinity Purified | |
RUO | |
27181 | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
For Research Use Only