missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse Siglec-2/CD22 Antibody, R&D Systems™

Goat Polyclonal Antibody
Brand: R&D Systems AF2296-SP
This item is not returnable.
View return policy
Description
Siglec-2/CD22 Polyclonal specifically detects Siglec-2/CD22 in Mouse samples. It is validated for Western Blot, Neutralization, Immunocytochemistry.
Specifications
| Siglec-2/CD22 | |
| Polyclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| B-cell receptor CD22, BL-CAM, B-lymphocyte cell adhesion molecule, CD22, CD22 antigenMGC130020, CD22 molecule, sialic acid binding Ig-like lectin 2, Sialic acid-binding Ig-like lectin 2, Siglec2, SIGLEC-2, SIGLEC2FLJ22814, T-cell surface antigen Leu-14 | |
| Goat | |
| Affinity Purified | |
| RUO | |
| 933 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
| Western Blot, Neutralization, Immunocytochemistry | |
| LYOPH | |
| Western Blot 0.1 ug/mL, Neutralization 0.1-0.5 ug/mL, Immunocytochemistry 5-15 ug/mL | |
| P35329 | |
| Cd22 | |
| Mouse myeloma cell line NS0-derived recombinant mouse Siglec-2/CD22 Ser22-Arg702 Accession # P35329 | |
| 25 μg | |
| Primary | |
| Detects mouse Siglec-2/CD22 in direct ELISAs and Western blots. In direct ELISAs, approximately 10% cross-reactivity with recombinant human (rh) Siglec-2 is observed. | |
| Mouse | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction