missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Mouse sFRP-3 ELISA Kit

Product Code. 18557774
Click to view available options
Quantity:
10 x 96 Tests
5 x 96 Tests
96 Tests
Unit Size:
Each
Pack of 10
Pack of 96
This item is not returnable. View return policy

Product Code. 18557774

Brand: Invitrogen™ EM69RBX10

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

ELISA

Mouse sFRP-3 quantitates mouse sFRP-3 in serum, plasma, supernatant. The assay will exclusively recognize both natural and recombinant mouse sFRP-3.

The protein encoded by this gene is a secreted protein that is involved in the regulation of bone development. Defects in this gene are a cause of female-specific osteoarthritis (OA) susceptibility.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number Fiz; Fre; FREOS1; frezzled; Fritz; frizzled homolog-related; frizzled-related protein; frizzled-related protein 1; FRP; FRP-3; FRZB; FRZB1; FrzB-1; FRZB1sFRP-3; FRZB-PEN; FZRB; hFIZ; OS1; Secreted frizzled-related protein 3; secreted frizzled-related sequence protein 3; Sfrp3; sFRP-3; SFRP3frezzled; SRFP3
Assay Range 25 to 6,000 pg/mL
Assay Sensitivity 25 pg/mL
Conjugate HRP
Product Type ELISA
Sample Type Plasma, Serum, Supernatant
For Use With (Equipment) Colorimetric Microplate Reader
Gene ID (Entrez) 20378
Gene Symbol FRZB
Interassay CV <12%
Intraassay CV <10
Kit Contents Pre-coated 96 well plate, Standard, Assay Diluent concentrate, Biotinylated Detection Antibody, SAV-HRP, Wash Buffer, Chromogen, Stop Solution, Adhesive Plate Covers
Label or Dye HRP
Quantity 10 x 96 Tests
Regulatory Status RUO
Sample Volume Plasma 50 μL, Serum 50 μL, Supernatant 50 μL
Storage Requirements 2°C to 8°C
Target FRZB
Test Time 1 hrs 20 mins
Total Assay Time 4 hrs 45 mins
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.