missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse sFRP-2 Antibody, R&D Systems™
Goat Polyclonal Antibody
Brand: R&D Systems AF1169
Additional Details : Weight : 0.00970kg
Description
sFRP-2 Polyclonal specifically detects sFRP-2 in Mouse samples. It is validated for Western Blot.Specifications
sFRP-2 | |
Polyclonal | |
Unconjugated | |
AAB70795 | |
SFRP2 | |
Mouse myeloma cell line NS0-derived recombinant mouse sFRP-2 (R&D Systems, Catalog # 1169-FR) Leu25-Cys295 Accession # AAB70795 | |
100 μg | |
Primary | |
Detects mouse sFRP-2 in direct ELISAs and Western blots. In these formats, less than 1% cross-reactivity with recombinant human SARP2 is observed. | |
Mouse | |
IgG |
Western Blot | |
LYOPH | |
Western Blot 0.1 ug/mL | |
FRP2, FRP-2secreted apoptosis related protein 1, SARP-1, SARP1sFRP-2, SDF-5, Secreted apoptosis-related protein 1, secreted frizzled-related protein 2, sFRP2 | |
Goat | |
Affinity Purified | |
RUO | |
6423 | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
For Research Use Only