missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse sFRP-2 Antibody, R&D Systems™

Rat Monoclonal Antibody
Brand: R&D Systems MAB1169-SP
This item is not returnable.
View return policy
Description
sFRP-2 Monoclonal specifically detects sFRP-2 in Mouse samples. It is validated for Western Blot.
Specifications
| sFRP-2 | |
| Monoclonal | |
| LYOPH | |
| Western Blot 1 ug/mL | |
| P97299 | |
| SFRP2 | |
| Mouse myeloma cell line NS0-derived recombinant mouse sFRP-2 Leu25-Cys295 Accession # P97299 | |
| 25 μg | |
| Primary | |
| Detects mouse sFRP-2 in direct ELISAs and Western blots. In direct ELISAs and Western blots, this antibody does not cross-react with recombinant human (rh) sFRP-1, recombinant mouse sFRP-3, or rhsFRP-4. | |
| Mouse | |
| Purified |
| Western Blot | |
| 331022 | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| FRP2, FRP-2secreted apoptosis related protein 1, SARP-1, SARP1sFRP-2, SDF-5, Secreted apoptosis-related protein 1, secreted frizzled-related protein 2, sFRP2 | |
| Rat | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 6423 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction