missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse Serpin E2/PN1 Antibody, R&D Systems™
Rat Monoclonal Antibody
Marca: R&D Systems MAB2175
Les retours ne sont pas autorisés pour ce produit.
Consulta la politica di reso
Descrizione
Serpin E2/PN1 Monoclonal specifically detects Serpin E2/PN1 in Mouse samples. It is validated for Western Blot, Immunoprecipitation.
Specifica
| Serpin E2/PN1 | |
| Monoclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| GDN, GdNPF, GDNserine (or cysteine) proteinase inhibitor, clade E (nexin, plasminogenactivator inhibitor type 1), member 2, glia-derived nexin, glial-derived neurite promoting factor, Peptidase inhibitor 7, PI-7, PI7DKFZp686A13110, PN-1, PN1nexin, PNI, Protease nexin 1, Protease nexin I, Proteinase Nexin 1, Serpin E2, serpin peptidase inhibitor, clade E (nexin, plasminogen activator inhibitortype 1), member 2 | |
| Rat | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 5270 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG2b |
| Western Blot, Immunoprecipitation | |
| 244404 | |
| Western Blot 1 ug/mL, Immunoprecipitation 25 ug/mL | |
| Q07235 | |
| SERPINE2 | |
| Mouse myeloma cell line NS0-derived recombinant mouse Serpin E2/PN1 Ser20-Pro397 Accession # Q07235 | |
| 500 μg | |
| Primary | |
| Detects mouse Serpin E2/PN1 in direct ELISAs and Western blots. In Western blots, no cross-reactivity with recombinant human Serpin A1, recombinant mouse (rm) Serpin D1, or rmSerpin F2 is observed. | |
| Mouse | |
| Purified |
Correzione del contenuto del prodotto
Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.
Titolo del prodotto
For Research Use Only
Individuate un'opportunità di miglioramento?Condividi una correzione di contenuto