missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse Serpin B8/Proteinase Inhibitor 8 Antibody, R&D Systems™

Goat Polyclonal Antibody
Brand: R&D Systems AF3588-SP
This item is not returnable.
View return policy
Description
Serpin B8/Proteinase Inhibitor 8 Polyclonal specifically detects Serpin B8/Proteinase Inhibitor 8 in Mouse samples. It is validated for Western Blot, Immunoprecipitation.
Specifications
| Serpin B8/Proteinase Inhibitor 8 | |
| Polyclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| CAP2, CAP-2, CAP2clade B (ovalbumin), member 8, Cytoplasmic antiproteinase 2, NK10, Peptidase inhibitor 8, PI8, PI-8, protease inhibitor 8 (ovalbumin type), serpin B8, serpin peptidase inhibitor, clade B (ovalbumin), member 8 | |
| Goat | |
| Affinity Purified | |
| RUO | |
| 5271 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
| Western Blot, Immunoprecipitation | |
| LYOPH | |
| Western Blot 0.1 ug/mL, Immunoprecipitation 25 ug/mL | |
| O08800 | |
| SERPINB8 | |
| S. frugiperda insect ovarian cell line Sf 21-derived recombinant mouse Serpin B8/Proteinase Inhibitor 8 Asp2-Pro374 Accession # O08800 | |
| 25 μg | |
| Primary | |
| Detects mouse Serpin B8/Proteinase Inhibitor 8 in direct ELISAs and Western blots. In Western blots, approximately 10% cross-reactivity with recombinant human (rh) Serpin B8, rhSerpin B9, and rhSerpin B6 is observed and less than 1% cross-reactivity with rhSerpin A1, D1, E2, I2, recombinant mouse Serpin A1, C1, E1, F1, and F2 is observed. | |
| Mouse | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction