missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse Semaphorin 4F Alexa Fluor 647-conjugated Antibody, R&D Systems™

Rat Monoclonal Antibody
Brand: R&D Systems FAB7200R-100UG
This item is not returnable.
View return policy
Description
Semaphorin 4F Monoclonal specifically detects Semaphorin 4F in Mouse samples. It is validated for Flow Cytometry.
Specifications
| Semaphorin 4F | |
| Monoclonal | |
| 0.2 mg/mL | |
| Flow Cytometry 0.25-1 ug/10^6 cells | |
| Q9Z123 | |
| SEMA4F | |
| Mouse myeloma cell line NS0-derived recombinant mouse Sema-4F Arg41-Gly667 Accession # Q9Z123 | |
| RUO | |
| 10505 | |
| Mouse | |
| IgG2a |
| Flow Cytometry | |
| 780225 | |
| Alexa Fluor 647 | |
| Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. with 0.09% Sodium Azide | |
| M-SEMA, m-Sema-M, PRO2353, sema domain, immunoglobulin domain (Ig), transmembrane domain (TM) and shortcytoplasmic domain, (semaphorin) 4F, sema domain, immunoglobulin domain (Ig), transmembrane domain (TM) and shortcytoplasmic domain, 4F, Sema M, Sema W, SEMA4F, SEMAM, semaphorin-4F, semaphorin-M, Semaphorin-W, SEMAW, SEMAWsemaphorin M | |
| Rat | |
| 100 μg | |
| Primary | |
| Detects mouse Sema-4F in ELISAs. | |
| Store the unopened product at 2 - 8 degreesC. Do not use past expiration date. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only (RUO)
Spot an opportunity for improvement?Share a Content Correction