missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse Semaphorin 3C Antibody, R&D Systems™

Sheep Polyclonal Antibody
Brand: R&D Systems AF1728-SP
This item is not returnable.
View return policy
Description
Semaphorin 3C Polyclonal specifically detects Semaphorin 3C in Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry.
Specifications
| Semaphorin 3C | |
| Polyclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| (semaphorin) 3C, sema domain, immunoglobulin domain (Ig), short basic domain, secreted, Sema E, SEMA3C, SEMAE, semaphorin E, semaphorin-3C, Semaphorin-E, SemE | |
| Sheep | |
| Affinity Purified | |
| RUO | |
| 10512 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
| Western Blot, Immunohistochemistry, Immunocytochemistry | |
| LYOPH | |
| Western Blot 0.1 ug/mL, Immunohistochemistry 5-15 ug/mL, Immunocytochemistry 5-15 ug/mL | |
| Q62181 | |
| SEMA3C | |
| Mouse myeloma cell line NS0-derived recombinant mouse Semaphorin 3C Gln24-Ser751 (Arg48Ala, Arg52Ala) Accession # Q62181 | |
| 25 μg | |
| Primary | |
| Detects mouse Semaphorin 3C in direct ELISAs and Western blots. In direct ELISAs, less than 5% cross-reactivity with recombinant human Semaphorin 3C, recombinant mouse (rm) Semaphorin 3A, rmSemaphorin 3B, rmSemaphorin 3E, and rmSemaphorin 3F is observed. | |
| Mouse | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction