missing translation for 'onlineSavingsMsg'
Learn More

Mouse S1P5/EDG-8 Antibody, R&D Systems™
GREENER_CHOICE

Rabbit Monoclonal Antibody

£178.00 - £386.00

Specifications

Antigen S1P5/EDG-8
Clone 1196A
Dilution Western Blot 1 ug/mL, Flow Cytometry 0.25 ug/10^6 cells
Applications Western Blot, Flow Cytometry
Classification Monoclonal
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
18764948
GREENER_CHOICE
View Documents
R&D Systems
MAB9084-SP
25 μg
£178.00
25µg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
18701309
GREENER_CHOICE
View Documents
R&D Systems
MAB9084
100 μg
£386.00
100µg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

S1P5/EDG-8 Monoclonal specifically detects S1P5/EDG-8 in Mouse samples. It is validated for Western Blot, Flow Cytometry.
Specifications

Specifications

S1P5/EDG-8
Western Blot 1 ug/mL, Flow Cytometry 0.25 ug/10^6 cells
Monoclonal
Purified
RUO
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with Sodium Azide
Edg-8, EDG8, EDG8S1P receptor Edg-8, Endothelial differentiation G-protein-coupled receptor 8, endothelial differentiation, sphingolipid G-protein-coupled receptor, 8, LPB4, S1P receptor 5, S1P5, S1PR5, sphingosine 1-phosphate receptor 5, sphingosine 1-phosphate receptor EDG8, Sphingosine 1-phosphate receptor Edg-8, sphingosine-1-phosphate receptor 5, SPPR-1, SPPR-2
S1pr5
IgG
Protein A or G purified from cell culture supernatant
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
1196A
Western Blot, Flow Cytometry
Unconjugated
Rabbit
Mouse
Q91X56
53637
Mouse S1P 5EDG-8 peptide corresponding to the N-terminal extracellular sequence Accession # Q91X56
Primary
Reconstitute at 0.5 mg/mL in sterile PBS.
Detects mouse S1P 5EDG-8 in Western blots.
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.