missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Mouse S100A8/S100A9 Heterodimer DuoSet ELISA
Click to view available options
Sufficient For:
Five 96-well plates
Unit Size:
Pack of 5
Description
The Mouse S100A8/S100A9 Heterodimer DuoSet ELISA is a Solid Phase Sandwich ELISA that quantifies mouse S100A8/S100A9 Heterodimer.
Specifications
Specifications
| Assay Range | 125 to 8,000pg/mL |
| Conjugate | Biotin |
| Immunoassay Kit Format | 96-well Strip Plate |
| Product Type | Solid Phase Sandwich ELISA |
| Sufficient For | Five 96-well plates |
| For Use With (Application) | ELISA |
| Storage Requirements | Store the unopened product at 2° to 8°C. Do not use past expiration date. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction