missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse S100A11 Antibody, R&D Systems™

Rat Monoclonal Antibody
Brand: R&D Systems MAB5167
This item is not returnable.
View return policy
Description
S100A11 Monoclonal specifically detects S100A11 in Mouse samples. It is validated for Western Blot.
Specifikationer
| S100A11 | |
| Monoclonal | |
| LYOPH | |
| Western Blot 1 ug/mL | |
| P50543 | |
| S100A11 | |
| E. coli-derived recombinant mouse S100A11 Met1-Ile98 Accession # P50543 | |
| 100 μg | |
| Primary | |
| Detects mouse S100A11 in direct ELISAs and Western blots. In direct ELISAs, ;no cross-reactivity with recombinant mouse S100A1, 4, 6, 8, 9, 10, 13, or 16 is observed, and approximately 75% cross-reactivity with recombinant human (rh) S100A11 is observed. In Western blots, no cross-reactivity with rhS100A11 is observed | |
| Mouse | |
| Purified |
| Western Blot | |
| 493804 | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| calgizzarin, Metastatic lymph node gene 70 protein, MLN 70, MLN70, protein S100-A11, Protein S100-C, S100 calcium binding protein A11, S100 calcium-binding protein A11, S100 calcium-binding protein A11 (calgizzarin), S100C, S100CS100 calcium binding protein A11 (calgizzarin) | |
| Rat | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 6282 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG2a |
Korrigering av produktinnehåll
Din input är viktig för oss. Fyll i det här formuläret för att ge feedback relaterad till innehållet på denna produkt.
Produkttitel
For Research Use Only
Hittar du en möjlighet till förbättring?Dela en innehållskorrigering