missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse Ret Antibody, R&D Systems™

Rat Monoclonal Antibody
Brand: R&D Systems MAB482-SP
This item is not returnable.
View return policy
Description
Ret Monoclonal specifically detects Ret in Mouse samples. It is validated for Western Blot.
Specifications
| Ret | |
| Monoclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| CDHF12MTC1, CDHR16, EC 2.7.10, Hirschsprung disease 1, Hirschsprung disease), MEN2A, MEN2B, multiple endocrine neoplasia and medullary thyroid carcinoma 1, ret proto-oncogene, RET transforming sequence | |
| Rat | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 5979 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG1 |
| Western Blot | |
| 64623 | |
| Western Blot 1 ug/mL | |
| P35546 | |
| RET | |
| Mouse myeloma cell line NS0-derived recombinant mouse Ret Leu29-Arg637 Accession # P35546 | |
| 25 μg | |
| Primary | |
| Detects mouse Ret in direct ELISAs and Western blots. In direct ELISAs and Western blots, this antibody shows 10-50% cross-reactivity with recombinant human Ret. | |
| Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction