missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse Reg2 Antibody, R&D Systems™
Goat Polyclonal Antibody
Brand: R&D Systems AF2035
Additional Details : Weight : 0.00970kg
Description
Reg2 Polyclonal specifically detects Reg2 in Mouse samples. It is validated for Western Blot, Immunohistochemistry.Specifications
Reg2 | |
Polyclonal | |
Unconjugated | |
Q08731 | |
Reg2 | |
E. coli-derived recombinant mouse Reg2 Asp35-Ala173 Accession # Q08731 | |
100 μg | |
Primary | |
Detects mouse Reg2 in direct ELISAs and Western blots. In Western blots, approximately 50% cross-reactivity with recombinant mouse Reg1 is observed and less than 1% cross-reactivity with recombinant human (rh) Reg1B, recombinant rat (rr) Reg2, rrReg3 and rhReg4 is observed. | |
Mouse | |
IgG |
Western Blot, Immunohistochemistry | |
LYOPH | |
Western Blot 0.1 ug/mL, Immunohistochemistry 5-15 ug/mL | |
regenerating islet-derived 2, RegII | |
Goat | |
Affinity Purified | |
RUO | |
5968 | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
For Research Use Only