missing translation for 'onlineSavingsMsg'
Learn More
Learn More
STEMCELL Technologies Mouse Recombinant SDF-1 alpha (CXCL12)
Click to view available options
Quantity:
25 μg
5 μg
Unit Size:
25µg
5µg
Description
- Stromal cell-derived factor 1 alpha (SDF-1α) is a member of the CXC group of chemokines that binds to the G-protein coupled receptor, CXCR4, to regulate migration, proliferation, differentiation, and survival of many cell types including hematopoietic stem cells, B cells, and T cells
- It is produced by bone marrow stromal cells, osteoblasts, endothelial cells, and neuronal cells
- SDF-1α was first identified as the pre-B-cell growth-stimulating factor (PBSF) in the mouse bone marrow-derived stromal cell line, PA6, in the growth of B cell precursors
- SDF-1α primarily regulates cell motility during development and adulthood, including the homing of hematopoietic stem cells and neutrophils to fetal bone marrow during ontogeny) and the recruitment of endothelial progenitor cells from bone marrow during angiogenesis in adulthood
Specifications
Specifications
| Name | CXCL12, PBSF, SDF-1 α, Stromal cell-derived factor-1 |
| Quantity | 25 μg |
| Cell Type | B Cells; Endothelial Cells; Epithelial Cells; Monocytes; T Cells |
| Purity or Quality Grade | ≥95% |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction