missing translation for 'onlineSavingsMsg'
Learn More
Learn More
STEMCELL Technologies Mouse Recombinant RANTES (CCL5)
Click to view available options
Quantity:
100 μg
20 μg
Unit Size:
100µg
20µg
Description
- RANTES (regulated upon activation, normal T cell expressed and secreted), also known as CCL5, is a member of the CC family of chemokines and is able to recruit leukocytes to sites of inflammation
- RANTES is secreted by T lymphocytes, macrophages, platelets, synovial fibroblasts, tubular epithelium, and certain types of tumor cells
- This chemokine exerts its effect by interacting with the chemokine receptors CCR1, CCR3, CCR4, and CCR5
- RANTES plays an active role in recruiting a variety of leukocytes into inflammatory sites, including T cells, macrophages, eosinophils, and basophils
- In collaboration with certain cytokines that are released by T cells such as IL-2 and IFN-γ, RANTES also induces the activation and proliferation of NK cells to generate CC chemokine-activated killer cells, which are highly cytolytic
Specifications
Specifications
| Name | D17S136, Eosinophil chemotactic cytokine, Regulated upon activation normally T-expressed and secreted, SIS-delta, Small-inducible cytokine A5, T-cell specific protein p288 |
| Quantity | 100 μg |
| Cell Type | B Cells; Dendritic Cells; Mesenchymal Stem and Progenitor Cells; Monocytes; NK Cells; T Cells; T Cells, CD4+; T Cells, CD8+ |
| Purity or Quality Grade | ≥95% |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction