missing translation for 'onlineSavingsMsg'
Learn More
Learn More
STEMCELL Technologies Mouse Recombinant IL-7
Click to view available options
Quantity:
10 μg
50 μg
Unit Size:
10µg
50µg
Description
- Interleukin 7 (IL-7) is a member of the type I cytokine family that is critical for T and B cell development and survival
- It is produced by non-hematopoietic cells in the thymus, lymphoid organs, and by bone marrow stromal cells (Lundström et al.) IL-7 binds to a receptor composed of common gamma chain and IL-7Ra (CD127) and signals through the JAK/STAT and PI3K pathways
- IL-7 regulates the survival of naïve and memory CD4+ and CD8+ T cells, γδ T cells, NK T cells, innate lymphoid cells, and T regulatory cells (Carrette & Surh), and provides signals to promote survival, proliferation, and differentiation during B cell development (Clark et al.; Corfe & Paige)
Specifications
Specifications
| Name | Interleukin-7, LP-1, Lymphopoietin 1, Pre-B cell factor |
| Quantity | 10 μg |
| Cell Type | Hematopoietic Stem and Progenitor Cells; Lymphocytes |
| Purity or Quality Grade | ≥95% |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction