missing translation for 'onlineSavingsMsg'
Learn More

STEMCELL Technologies Mouse Recombinant IL-17A

Product Code. p-7217723
Encompass_Preferred
Click to view available options
Quantity:
100 μg
25 μg
Unit Size:
100µg
25µg
This item is not returnable. View return policy

Product Code. 17159401

Brand: STEMCELL Technologies 78033

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Interleukin 17A

  • Interleukin 17A (IL-17A) is the founding member of the family of cytokines that includes Interleukin 17B through Interleukin 17F
  • It is a potent proinflammatory cytokine that plays a key role in defense against pathogens
  • IL-17A and IL-17F signal as homodimers or heterodimers through the same receptor, and activate NF-kB, MAPK, and C/EBP pathways
  • IL-17A receptor is expressed on a variety of cell types, including hematopoietic cell compartments
  • IL-17A is produced by T helper 17 cells, CD8+ T cells, γδ T cells, natural killer T cells, B cells, neutrophils, innate lymphoid cells and mesenchymal stromal cells (MSCs)
  • IL-17A receptor is expressed at particularly high levels on stromal cells, including MSCs
  • IL-17A increases the frequency and the average size of colony-forming units-fibroblast derived from bone marrow, as well as the proliferation of bone marrow-derived MSCs

Specifications

Name CTLA-8, IL-17, Interleukin-17, Interleukin-17A
Quantity 100 μg
Cell Type B Cells; Lymphocytes; T Cells; T Cells, CD4+; T Cells, CD8+
Purity or Quality Grade ≥95%
Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.